Stampa…

Anti-IgG (H+L) Goat Polyclonal Antibody (Agarose)

Fornitore: Rockland Immunochemicals
201-1150
ROCK201-1150EA 453 EUR
ROCK201-1150
Anti-IgG (H+L) Goat Polyclonal Antibody (Agarose)
Anticorpi
Anti-Bovine IgG (H&L) (GOAT) Antibody Agarose Conjugated is applicable to purification of Bovine IgG from sample solution.

Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Agarose
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Bovine
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR