Stampa…

Anti-UBB Rabbit Polyclonal Antibody

Fornitore: Rockland Immunochemicals
200-401-431
ROCK200-401-431EA 906 EUR
ROCK200-401-431
Anti-UBB Rabbit Polyclonal Antibody
Anticorpi
This purified polyclonal antibody reacts with ubiquitin by ELISA and western blot. Although not tested, this antibody is likely functional in immunochemistry and immunoprecipitation. For detection of free Ub by western blotting use Tris-Tricine SDS-PAGE.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:200 - 1:1000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: UBB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pan
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR