Stampa…

Anti-HSPD1 Mouse Monoclonal Antibody (Unconjugated) [clone: LK2]

Fornitore: Rockland Immunochemicals
200-301-F61
ROCK200-301-F61EA 841 EUR
ROCK200-301-F61
Anti-HSPD1 Mouse Monoclonal Antibody (Unconjugated) [clone: LK2]
Anticorpi
Anti-Hsp60 Antibody is suitable for WB, Flow Cytometry, and IHC. Expect a band approximately ~60kDa Protein corresponding to the molecular mass of Hsp60 on SDS PAGE immunoblots.

Recommended Dilutions: Western Blot: 0.25 µg/ml;

Type: Primary
Antigen: HSPD1
Clonality: Monoclonal
Clone: LK2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Bovine, Dog, Chicken, Guinea Pig, Hamster, Monkey, Pig, Rabbit, Spinach, E. coli, H. Pylori, S. Typhimurium, T. Spiralis , Yeast, White Fly
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR