Stampa…

Anti-YDJ1 Mouse Monoclonal Antibody (Unconjugated) [clone: 1G10.H8]

Fornitore: Rockland Immunochemicals
200-301-E58
ROCK200-301-E58EA 841 EUR
ROCK200-301-E58
Anti-YDJ1 Mouse Monoclonal Antibody (Unconjugated) [clone: 1G10.H8]
Anticorpi
Anti-Hsp40 Antibody is recommended for use in WB, IP, and ELISA. Expect a band approximately ~40kDa Protein corresponding to the molecular mass of Hsp40 on SDS PAGE immunoblots. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1:2000;

Type: Primary
Antigen: YDJ1
Clonality: Monoclonal
Clone: 1G10.H8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Yeast
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR