Stampa…
Filter discs, biplane, series 16, porosity 3, VitraPOR®
Fornitore: ROBU GLASFILTER
16303 16403 16503 16603 16703 16803 16903 16113 16123 16103 16203 16253 61803 62003 61903 61303 62303 62703 62503 62403 62603 63803 62803 63503 63003 62903 61703 61603 61503 61403 62203 62103
ROBU16303EA 82.4 EUR
ROBU16303 ROBU16403 ROBU16503 ROBU16603 ROBU16703 ROBU16803 ROBU16903 ROBU16113 ROBU16123 ROBU16103 ROBU16203 ROBU16253 ROBU61803 ROBU62003 ROBU61903 ROBU61303 ROBU62303 ROBU62703 ROBU62503 ROBU62403 ROBU62603 ROBU63803 ROBU62803 ROBU63503 ROBU63003 ROBU62903 ROBU61703 ROBU61603 ROBU61503 ROBU61403 ROBU62203 ROBU62103
Filter discs, biplane, series 16, porosity 3, VitraPOR®
Filtri Dischi filtranti
The unique porous structure of VitraPOR® sintered glass filters makes it flexible and versatile for many applications.
  • Very good chemical resistance
  • Very good temperature resistance
Made of borosilicate glass. The faces of filter discs from the series 16 are precisely ground down to tight tolerances and plane surfaces as may be required in specific installation circumstances.