Stampa…
Filter discs, biplane, series 16, porosity 5, VitraPOR®
Fornitore: ROBU GLASFILTER
16105 16115 16125 16205 16255 16305 16405 16505 16605 16705 16805 16905
ROBU16105EA 176 EUR
ROBU16105 ROBU16115 ROBU16125 ROBU16205 ROBU16255 ROBU16305 ROBU16405 ROBU16505 ROBU16605 ROBU16705 ROBU16805 ROBU16905
Filter discs, biplane, series 16, porosity 5, VitraPOR®
Filtri Dischi filtranti
The unique porous structure of VitraPOR® sintered glass filters makes it flexible and versatile for many applications.
  • Very good chemical resistance
  • Very good temperature resistance
Made of borosilicate glass. The faces of filter discs from the series 16 are precisely ground down to tight tolerances and plane surfaces as may be required in specific installation circumstances.