Stampa…

Piombo acetato su cotone Reag. Ph. Eur. 1048101

Fornitore: REAGECON

Synonyms: Lead (II) acetate on cotton, Lead di-acetate

1048101
REGE1048101EA 285 EUR
REGE1048101
Piombo acetato su cotone Reag. Ph. Eur. 1048101
Piombo acetato su cotone

  • Composition: 1 dilute acetic acid:10 lead acetate solution
  • Form: Cottony balls


Manufactured according to European Pharmacopoeia Chapter 4 (4.1.1) and prepared in an ISO 9001 facility. Lead Acetate Cotton, EP Grade, Reagecon™ is a ready-to-use, dried cotton that has absorbed a 10:1 lead acetate-dilute acetic acid solution for use in arsenic limit tests.

Formula: (H₃CCOO)₂Pb
Peso molecolare: 325.29 g/mol
CAS Number: 6080-56-4
UN: 3077
ADR: 9 III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR