Stampa…
Disposable particle filtering respirators, FFP2, P251
Fornitore: Portwest
P251WHR
PRTWP251WHREA 25.9 EUR
PRTWP251WHR
Disposable particle filtering respirators, FFP2, P251
Respiratori Maschere monouso Respiratori antiparticolato
Vertical flat folded mask with exhalation valve for lighter breathing resistance. Ideal for all day use.
  • Adjustable nose clip for optimised fit
  • Ultrasound welded elastic bands
  • Flexible shape allows the mask to be folded flat
  • Individually packed for vending machines
Certificazioni: EN 149:2001 + A1:2009.
Confezione: Individually wrapped inside hygienic polybag.