Stampa…
Elemental analysis standards, PTP sample of oils, VHG
Fornitore: LGC Standards PROMOCHEM
VHG-VPTPMO-25G
PROOVHG-VPTPMO-25GEA 147 EUR
PROOVHG-VPTPMO-25G
Elemental analysis standards, PTP sample of oils, VHG
Standard Standard per l'industria peltrolchimica
The PTP has been specially designed for analytical laboratories that analyse new and used oils for metals, sulphur, viscosity and particle count.
  • Monitor and improve measurements processes
  • Demonstrate analytical competency
  • Method and instrumentation validation
  • 25 analytes
Certificazioni: ISO/IEC 17043 accredited.