Stampa…
Arsenic standard solutions, VHG
Fornitore: LGC Standards PROMOCHEM
VHG-SPAS3-100 VHG-SPAS5W-100 VHG-SPAS3-50 VHG-SPAS5W-50
PROOVHG-SPAS3-100EA 51 EUR
PROOVHG-SPAS3-100 PROOVHG-SPAS5W-100 PROOVHG-SPAS3-50 PROOVHG-SPAS5W-50
Arsenic standard solutions, VHG
Standard Standard cationici Standard di arsenico
This standard solution is intended for use as a calibration or reference standard for plasma emission spectroscopy (ICP or DCP), inductively coupled plasma mass spectroscopy (ICP-MS), atomic absorption spectroscopy (AA or GFAA) or techniques using other modes of elemental detection and an appropriate separation method.
Certificazioni: Accredited to ISO 9001 and ISO/IEC 17025. Traceable to NIST and SRM.