Stampa…
Chromium standard solution, for ICP, in 5% HNO₃, VHG
Fornitore: LGC Standards PROMOCHEM
VHG-PCRN-100 VHG-PCRN-500 VHG-PCRN-50 VHG-TCRN-100 VHG-TCRN-500
PROOVHG-PCRN-100EA 50 EUR
PROOVHG-PCRN-100 PROOVHG-PCRN-500 PROOVHG-PCRN-50 PROOVHG-TCRN-100 PROOVHG-TCRN-500
Chromium standard solution, for ICP, in 5% HNO₃, VHG
Standard Standard cationici Standard di cromo
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificazioni: Traceable to NIST, SRM.