Stampa…
DNPH standards, organotin mixtures, CERTAN®
Fornitore: LGC Standards PROMOCHEM
SL31005
PROOSL31005EA 607 EUR
PROOSL31005
DNPH standards, organotin mixtures, CERTAN®
Standard Standard organici
These selected organotin compounds are determined by gas chromatography using DIN 38407-F13.
Certificazioni: According to DIN EN ISO 17353.