Stampa…
Iron standard solution
Fornitore: LGC Standards PROMOCHEM
NIST-3126A
PROONIST-3126AEA 1170 EUR
PROONIST-3126A
Iron standard solution
Standard Standard cationici Standard di ferro
This standard reference material is intended for use as a primary calibration standard for the quantitative determination of iron.
A unit consists of 50 ml of a single element solution in a high density polyethylene bottle sealed in an aluminised bag.
Certificazioni: Traceable to NIST and SRM numbers.