Stampa…

Dexamethasone

Fornitore: LGC Standards PROMOCHEM

ICRS0267
PROOICRS0267EA 226 EUR
PROOICRS0267
Dexamethasone
Dexamethasone
Formula: C₂₂H₂₉FO₅
Punto di fusione: 268…271 °C
MDL Number: MFCD00064136
CAS Number: 50-02-2
EINECS: 200-003-9

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR