Stampa…
Reference standards, biodiesel/diesel fuel Blends
Fornitore: LGC Standards PROMOCHEM
GBDBLEND10P-20 GBDBLEND10P-50 GBDBLEND15P-20 GBDBLEND20P-20 GBDBLEND25P-20 GBDBLEND-2P-20 GBDBLEND30P-20 GBDBLEND-5P-20 GBDBLEND5P-250 GBDBLEND-5P-50 GBDBLENDBLK-20
PROOGBDBLEND10P-20EA 96 EUR
PROOGBDBLEND10P-20 PROOGBDBLEND10P-50 PROOGBDBLEND15P-20 PROOGBDBLEND20P-20 PROOGBDBLEND25P-20 PROOGBDBLEND-2P-20 PROOGBDBLEND30P-20 PROOGBDBLEND-5P-20 PROOGBDBLEND5P-250 PROOGBDBLEND-5P-50 PROOGBDBLENDBLK-20
Reference standards, biodiesel/diesel fuel Blends
Standard Standard per l'industria peltrolchimica
These standards are intended for use as calibration and reference standards for the determination of fatty acid methyl esters (FAME) biodiesel content in diesel fuel by infrared (IR) spectroscopy. They are formulated specifically for use with ASTM Method D7371 and EN 14078.