Stampa…

2,3,3',4',5'-Pentachlorobiphenyl (PCB No. 122) 35 µg/ml in isooctane

Fornitore: LGC Standards PROMOCHEM

CIL-PCB-122
PROOCIL-PCB-122EA 150 EUR
PROOCIL-PCB-122
2,3,3',4',5'-Pentachlorobiphenyl (PCB No. 122) 35 µg/ml in isooctane
Standard Standard organici

Visualizza i codici

Specifiche

Descrizione 2,3,3',4',5'-Pentachlorobiphenyl (PCB No. 122)
Environmentally Preferable
CAS No. 76842-07-4
Formula C₁₂H₅Cl₅

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR