Stampa…

Organic reference standards, Azapropazone assay standard, BP

Fornitore: LGC Standards PROMOCHEM

BP534
PROOBP534EA 297 EUR
PROOBP534
Organic reference standards, Azapropazone assay standard, BP
Standard Standard organici

Visualizza i codici

Specifiche

Descrizione Azapropazone assay standard
Environmentally Preferable
CAS No. 13539-59-8
Formula C₁₆H₂₀N₄O₂

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR