Stampa…
Cleaning trolley Clino®, Profi C5e
Fornitore: PFENNIG REINIGUNGSTECHNIK
2095520
PPSA2095520EA 1530 EUR
PPSA2095520
Cleaning trolley Clino®, Profi C5e
Carrelli
The Clino® Profi C5e trolley system made of stainless steel flat wringer and three buckets for surface cleaning.
  • Minimal footprint
  • Highest stability through construction of tubular frame
  • Flat wringer Clino provides defined pressure point for uniform residual moisture
  • Double ball-bearinged guide wheels (125 mm)
Informazioni per l'ordine: Supplied with buckets (25 L, 15 L and 3 ×6 L) and disposal unit (2 ×70 L).