Stampa…
Membrane protein extraction kit, Pierce™ Mem-PER™ Plus
Fornitore: Thermo Fisher Scientific
89842
PIER89842EA 473 EUR
PIER89842
Membrane protein extraction kit, Pierce™ Mem-PER™ Plus
Estrazione delle proteine Kit per l'estrazione delle proteine
The Mem-PER™ Plus Membrane Protein Extraction Kit enables small-scale solubilisation and enrichment of integral membrane proteins and membrane-associated proteins in a simple reagent-based procedure. It effectively isolates membrane proteins from cultured cells and tissues using a simple benchtop microcentrifuge procedure.
  • Produces minimal cross-contamination (typically less than 10%) of cytosolic protein into the membrane protein fraction
  • Effective for extraction from cultured mammalian cells and mammalian tissues
  • Suitable for downstream applications like SDS-PAGE, Western blotting, immunoprecipitation and protein assays
  • Fast and simple with isolation of membrane proteins in approximately one hour
  • No special equipments required, only a benchtop microcentrifuge, tubes, homogenisers, and pipettors are needed
The Mem-PER™ Plus Membrane Protein Extraction Kit makes possible the extraction and selective enrichment of integral and attached membrane proteins from cultured mammalian cells, as well as from hard and soft mammalian tissues.
Informazioni per l'ordine: The kit contains a cell wash solution (225 ml), permeabilisation buffer (50 ml), and solubilisation buffer (25 ml).