Stampa…
Chemiluminescence exposure screen for myECL™ imager
Fornitore: Thermo Fisher Scientific
62242
PIER62242EA 698 EUR
PIER62242
Chemiluminescence exposure screen for myECL™ imager
Sistemi di documentazione fotografica
This black plastic sample screen is used to cover the transilluminator on the myECL™ imager.
This has been used as a replacement for the accessory used on the transilluminator for acquiring chemiluminescent images.