Stampa…
StartingBlock™ Blocking Buffer
Fornitore: Thermo Fisher Scientific
37542 37538 37539 37543 37579 37578
PIER37542EA 0 EUR
PIER37542 PIER37538 PIER37539 PIER37543 PIER37579 PIER37578
StartingBlock™ Blocking Buffer
Soluzioni tampone Tamponi per elettroforesi Tamponi di bloccaggio
These StartingBlock™ blocking buffers are PBS and TBS formulations of a proprietary protein that provides exceptional performance and broad compatibility with membrane (Western blot) and 96-well plate (ELISA) assays.
  • Compatible with many detection systems (Western blot, ELISA and IHC)
  • Short blocking times
  • High signal:noise ratio
  • Strip and reprobe without reblocking
  • Available ready to use 1X formulations in PBS and TBS
  • For research use only
  • Not for use in diagnostic procedures
It provides broad compatibility with a wide range of antibodies, antibody combinations, and other protein probing and assay systems. StartingBlock blocking buffer is available in PBS and TBS, with and without Tween 20.