Stampa…
Manganese standard solution, for ICP-MS, Pure Plus grade
Fornitore: PerkinElmer
N9303744
PERCN9303744EA 105 EUR
PERCN9303744
Manganese standard solution, for ICP-MS, Pure Plus grade
Standard Standard cationici Standard di manganese
Single element standard is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
  • Analysed by ICP-OES
  • Analysed by classical wet assay
  • 67 trace impurities analysed by ICP-MS of the final solution and reported on the certificate
  • Impurities reported at ppb level
  • Prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Attenzione: These standards are in acid solution, for full details of hazards please read the accompanying Material Safety Data Sheets (MSDS).