Stampa…

Anti-Calbindin D-28K (Ab-1) Rabbit Polyclonal Antibody

Fornitore: Merck Millipore (Oncogene)
PC253L-100
PC253L-100EA 905 EUR
PC253L-100
Anti-Calbindin D-28K (Ab-1) Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: Calbindin D-28K (Ab-1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR