Stampa…
Centrifugal devices for RNA and DNA extraction, NAB Nanosep®
Fornitore: Cytiva (Formerly Pall Lab)
FD001X34 ODNABC33 ODNABC34
516-9341EA 152 EUR
516-9341 PALLODNABC33 PALLODNABC34
Centrifugal devices for RNA and DNA extraction, NAB Nanosep®
Filtri Filtri centrifughi
The Pall Nanosep device for nucleic acid binding (NAB) incorporates a dual layer silica-based quartz glass fiber media to allow for efficient binding of DNA and RNA, while providing smooth flow and rapid processing of samples.
  • Rapid processing of samples
  • Silica-based quartz glass fiber media that allows efficient binding of DNA and RNA
  • Constructed of low-binding polypropylene
  • Ultrasonically welded seals prevent by passor seal failure
  • Fits standard centrifuge rotors that accept 1,5 ml tubes
This media offers researchers the flexibility to purifyMore Product Information