Stampa…

Glicerina Ph. Eur., Macron Fine Chemicals™

Fornitore: Avantor Performance Materials

P137-33EA 70.05 EUR
P137-33
Glicerina Ph. Eur., Macron Fine Chemicals™
Glicerina
Formula: HOCH₂CH(OH)CH₂OH
MDL Number: MFCD00004722
CAS Number: 56-81-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR