Stampa…

Acido cloridrico soluzione 10% Biotech Reagent, meets analytical specification of Ph.Eur., Macron Fine Chemicals™

Fornitore: Avantor Performance Materials

P111-33EA 29.54 EUR
P111-33 P111-38 P111-39 P111-40
Acido cloridrico soluzione 10% Biotech Reagent, meets analytical specification of Ph.Eur., Macron Fine Chemicals™
Acido cloridrico
Formula: HCl
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR