Stampa…
Culti-Loops™ Streptococcus agalactiae ATCC™ 12401™
Fornitore: THERMO FISHER DIAGNOSTICS
R4609453
OXOIR4609453EA 262.5 EUR
OXOIR4609453
Culti-Loops™ Streptococcus agalactiae ATCC™ 12401™
Test per microbiologia Test sui microrganismi
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Confezione: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.