Stampa…
Culti-Loops™ Streptococcus pneumoniae ATCC ™6303™
Fornitore: THERMO FISHER DIAGNOSTICS
R4608221
OXOIR4608221EA 262.5 EUR
OXOIR4608221
Culti-Loops™ Streptococcus pneumoniae ATCC ™6303™
Test per microbiologia Test sui microrganismi
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Confezione: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.