Stampa…
Culti-Loops™ Gemella morbillorum ATCC™ 27824™
Fornitore: THERMO FISHER DIAGNOSTICS
R4602100
OXOIR4602100EA 262.5 EUR
OXOIR4602100
Culti-Loops™ Gemella morbillorum ATCC™ 27824™
Test per microbiologia Test sui microrganismi
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, or streaked directly onto an appropriate media.
Confezione: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.