Stampa…
Culti-Loops™ Aspergillus oryzae ATCC™ 10124™
Fornitore: THERMO FISHER DIAGNOSTICS
R4601017
OXOIR4601017EA 262.5 EUR
OXOIR4601017
Culti-Loops™ Aspergillus oryzae ATCC™ 10124™
Test per microbiologia Test sui microrganismi
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Confezione: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.