Stampa…
Dip slides
Fornitore: THERMO FISHER DIAGNOSTICS
DS0147A DS0155A DS0166A DS0168A DS0169A DS0170A GFG04A GFD04A GFD02A GFD03A GFD05A GFD06A GFD07A
OXOIDS0147AEA 53.9 EUR
OXOIDS0147A OXOIDS0155A OXOIDS0166A OXOIDS0168A OXOIDS0169A OXOIDS0170A OXOIGFG04A OXOIGFD04A OXOIGFD02A OXOIGFD03A OXOIGFD05A OXOIGFD06A OXOIGFD07A
Dip slides
Terreni per microbiologia Terreni pronti per microbiologia
Simple and safe for an unskilled operator to use, these dip slides are coated on both sides with a solid culture medium with the opportunity for different media to be carried on each side, making the process time- and cost-effective. This sterile dip slide can be inoculated in the ward or doctor’s surgery and then returned to its plastic container for easy transport back to the laboratory.
  • Make colony counting for the bacterial assessment simple