Stampa…
Culture media in bags, ReadyBags™
Fornitore: THERMO FISHER DIAGNOSTICS
BM0104T BM0204T BM1104T BM9901A BP0100C BP0100E BP0100K BP0100R
OXOIBM0104TEA 230 EUR
OXOIBM0104T OXOIBM0204T OXOIBM1104T OXOIBM9901A OXOIBP0100C OXOIBP0100E OXOIBP0100K OXOIBP0100R
Culture media in bags, ReadyBags™
Terreni per microbiologia Terreni pronti per microbiologia
ReadyBags™ are designed for use in food testing by laboratories with large numbers of samples requiring large volumes of broth media or diluent. Each ReadyBag™ contains 3 liters and is intended for use with an automatic dispenser. The ReadyBag™ is connected to tubing for standard laboratory equipment, such as gravimetric diluters or peristaltic pumps, by means of a connector. Connectors are available in two formats: autoclavable, re-usable stainless steel connectors or sterile, plastic, single-use connectors.
  • For large numbers of samples
  • Available in 3 L
  • Once empty, the bag can be disposed as normal laboratory waste