Stampa…
SFB expansion
Fornitore: Oxford Nanopore Technologies
EXP-SFB001
OXNTEXP-SFB001EA 30 EUR
OXNTEXP-SFB001
SFB expansion
Reagenti per acidi nucleici Reagenti di sequenziamento di nuova generazione
Short Fragment Buffer (SFB).
This kit is recommended for users who require additional Short Fragment Buffer for use with the 'PCR tiling of COVID-19 virus' protocol or any other experiment sequencing short fragments.
SFB Expansion contains four vials of Short Fragment Buffer (SFB), sufficient for four reactions.
Informazioni per l'ordine: Flow cells and kits are shipped together at 2 to 8 °C. Upon receipt, please place the product in a long-term storage location at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be three months from receipt by the customer.
Attenzione: Oxford Nanopore Technologies products are currently for research use only.
Visualizza i codici
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.