Stampa…
Filling System, RF-3
Fornitore: THERMO OWL SCIENTIFIC
RF-3
OWLARF-3EA 384 EUR
OWLARF-3
Filling System, RF-3
Arredi Ripiani
Organize your refrigerator, coldroom, freezer or benchtop with the Thermo Scientific™ Owl™ RF-3 Filing System.
  • Eight compartments constructed of durable 640 mm thick acrylic
  • No-skid rubber feet prevent sliding
  • Use either vertically or horizontally