Stampa…
Silver billet electrode, Orion™
Fornitore: Thermo Orion
9780SC
ORIO9780SCEA 287 EUR
ORIO9780SC
Silver billet electrode, Orion™
Elettrodi Altri elettrodi
These electrodes are a combination of a silver billet-indicating electrode along with a silver billet half-cell. Ideal for obtaining an accurate determination of potentiometric halides with dual electrodes.
Internal reference: Ag/AgCl