Stampa…
Human Recombinant TEX264 (NM_015926)
Fornitore: OriGene
TP300008M
ORIGTP300008MEA 4100 EUR
ORIGTP300008M
Human Recombinant TEX264 (NM_015926)
Proteine e Peptidi
Recombinant protein of human testis expressed 264 (TEX264), transcript variant 1, 100 µg.
Major reticulophagy (also called ER-phagy) receptor that acts independently of other candidate reticulophagy receptors to remodel subdomains of the endoplasmic reticulum into autophagosomes upon nutrient stress, which then fuse with lysosomes for endoplasmic reticulum turnover. The ATG8-containing isolation membrane (IM) cradles a tubular segment of TEX264-positive ER near a three-way junction, allowing the formation of a synapse of two juxtaposed membranes with trans interaction between the TEX264 and ATG8 proteins. Expansion of the IM would extend the capture of ER, possibly through a 'zipper-like' process involving continued trans TEX264-ATG8 interactions, until poorly understood mechanisms lead to the fission of relevant membranes and, ultimately, autophagosomal membrane closure.
Protein Families: Secreted Protein, Transmembrane