Stampa…

Anti-DPYSL4 Rabbit Polyclonal Antibody

Fornitore: OriGene
TA322499
ORIGTA322499EA 528 EUR
ORIGTA322499
Anti-DPYSL4 Rabbit Polyclonal Antibody
Anticorpi
Recommended Dilutions: ELISA: 1:1000-5000; Western Blot: 1:500-1000; Immunochemistry: 1:25-100

Type: Primary
Antigen: DPYSL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR