Stampa…

Anti-p53 (Ab-5) (Wild type) (PAb1620) Mouse Monoclonal Antibody [clone: PAb1620]

Fornitore: Merck Millipore (Oncogene)
OP33-100
OP33-100EA 755 EUR
OP33-100
Anti-p53 (Ab-5) (Wild type) (PAb1620) Mouse Monoclonal Antibody [clone: PAb1620]
Anticorpi
Type: Primary
Antigen: p53 (Ab-5) (Wild type) (PAb1620)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR