Stampa…

Anti-c-ErbB2/c-Neu (Ab-2) (9G6) Mouse Monoclonal Antibody [clone: 9G6]

Fornitore: Merck Millipore (Oncogene)
OP14-100
OP14-100EA 667 EUR
OP14-100
Anti-c-ErbB2/c-Neu (Ab-2) (9G6) Mouse Monoclonal Antibody [clone: 9G6]
Anticorpi
Type: Primary
Antigen: c-ErbB2/c-Neu (Ab-2) (9G6)
Clonality: monoclonal
Clone: 9G6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR