Stampa…

Anti-p53 (Ab-11) (Pantropic) (PAb1802) Mouse Monoclonal Antibody

Fornitore: Merck Millipore (Oncogene)
OP104L-100
OP104L-100EA 740 EUR
OP104L-100
Anti-p53 (Ab-11) (Pantropic) (PAb1802) Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: p53 (Ab-11) (Pantropic) (PAb1802)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR