Stampa…
Caps for solvent safety bottles, T-Series
Fornitore: OMNIFIT
00945T-2 00932T-2 00945T-2F 00945T-3 00945T-3F 00945T-4 00945T-4F
OMNI00945T-2EA 69.3 EUR
OMNI00945T-2 OMNI00932T-2 OMNI00945T-2F OMNI00945T-3 OMNI00945T-3F OMNI00945T-4 OMNI00945T-4F
Caps for solvent safety bottles, T-Series
Ghiere Tappi per bottiglie
Outer PP cap, inner PTFE body.
  • Simple and easy-to-use with no need for fitting nuts and ferrules (for ¹/₈" Ext. Ø semi-rigid tube). Tubing passes straight through the cap for a continuous fluid path
  • Luer ports take standard male Luer components including a range of accessories for different tubing types and sizes
  • Optional integrated check valve and filter enables pressure equalisation while keeping the system closed and preventing particulate contamination or evaporation of expensive or harmful chemicals
  • A Luer connection solvent vapor trap can be used to