Stampa…
Reciprocating shaker
Fornitore: OHAUS
30391830
OHAU30391830EA 2340 EUR
OHAU30391830
Reciprocating shaker
Agitatori Agitatori con moto alternato
This digital reciprocating shaker is designed for a wide range of applications, including cell cultures, that require accurate and repeatable results. The microprocessor control provides consistent uniform shaking while safely ramping to the set speed. Ideal for cell cultures, solubility studies and extraction procedures.
  • Microprocessor will display last set-point and will restart if power is interrupted
  • RS232 interface provides two-way communication for data logging and unit control
  • Speed calibration mode allows user to automatically recalibrate speed
More Product Information
Visualizza i codici
Specifiche
Modello SHRC0719DG
Environmentally Preferable
Movimento di agitazione Reciprocating
Corse 19 mm
Intervallo di velocità 20 - 300 min⁻¹
Potenza 40 W
Timer 1 s – 160 h
L×P piattaforma 279×330 mm
L×P×A 413×356×146 mm
Peso 21,8 kg
Carico massimo 6,8 kg
Elettrico 230 V, 2,5 A, 50/60 Hz