Stampa…

Anti-LGR5 Rabbit Polyclonal Antibody (Biotin)

Fornitore: Novus Biologicals
NLS1235B
NOVUNLS1235BEA 704 EUR
NOVUNLS1235B
Anti-LGR5 Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: LGR5
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR