Stampa…

Anti-p27/Kip1 Mouse Monoclonal Antibody [clone: SPM348]

Fornitore: Novus Biologicals
NBP2-32890-0.1
NOVUNBP2-32890-0.1EA 583 EUR
NOVUNBP2-32890-0.1
Anti-p27/Kip1 Mouse Monoclonal Antibody [clone: SPM348]
Anticorpi
p27(Kip1) is a cyclin-dependent kinase inhibitor involved in G1 arrest. p27(Kip1) binds to and inhibits cyclinE-Cdk2 complex, cyclinA-CDK2 and cyclinD1-CDK4 (1). p27(Kip1) is regulated by phosphorylation on serine 10 (S10) and threonine 187 (T187). Phosphorylation by CDK2 on T187 results in ubiquitylation and degradation of p27(Kip1) (2); while phosphorylation by hKIS on S10 signals the nuclear export to the cytoplasm (3).
Visualizza i codici

Specifiche

Simbolo dell'antigene P27/KIP
Environmentally Preferable
Tipo di anticorpi Cyclin-dependent kinase inhibitor p27
Sinomini antigene CDKN1B|CDKN4
Clonalità Monoclonale
Coniugazione Non coniugato
Clone SPM348
Reattività Primates,Human,Rat,Mouse
Ospite Mouse
Isotipo IgG1 kappa
ImmunoChimica Yes
ImmunoFluorescenza Yes
Immunogeno Purified GST-p27 fusion protein of Mouse origin
Purificazione Protein A or G purified
Concentrazione 0.2 mg/ml
Tampone di conservazione 10 mM PBS with 0.05% BSA; Preservative:
0.05% Sodium Azide
Temperatura di conservazione Store at 4 °C
UniProtKB P46527

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR