Stampa…

Anti-MAP2K1 Rabbit Polyclonal Antibody

Fornitore: Novus Biologicals
NBP2-24542
NOVUNBP2-24542EA 535 EUR
NOVUNBP2-24542
Anti-MAP2K1 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: MAP2K1 (mitogen-activated protein kinase kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR