Stampa…

Anti-DSG3 Mouse Monoclonal Antibody (DyLight® 350) [clone: 5H10]

Fornitore: Novus Biologicals
NBP1-78984UV
NOVUNBP1-78984UVEA 668 EUR
NOVUNBP1-78984UV
Anti-DSG3 Mouse Monoclonal Antibody (DyLight® 350) [clone: 5H10]
Anticorpi
Type: Primary
Antigen: DSG3 (desmoglein 3)
Clonality: monoclonal
Clone: 5H10
Conjugation: DyLight® 350
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR