Stampa…

Anti-HMOX1 Rabbit Polyclonal Antibody

Fornitore: Novus Biologicals
NBP1-77460 NBP1-77460SS
NOVUNBP1-77460EA 554 EUR
NOVUNBP1-77460 NOVUNBP1-77460SS
Anti-HMOX1 Rabbit Polyclonal Antibody
Anticorpi
Rabbit Polyclonal Heme Oxygenase 1 Antibody. Tested Applications: Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: HMOX1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR