Stampa…

Anti-DUSP10 rabbit polyclonal antibody

Fornitore: Novus Biologicals
NBP1-31001
NOVUNBP1-31001EA 692 EUR
NOVUNBP1-31001
Anti-DUSP10 rabbit polyclonal antibody
Anticorpi
The DUSP10 Antibody from Novus Biologicals is a rabbit polyclonal antibody to DUSP10. This antibody reacts with human. The DUSP10 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: DUSP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR