Stampa…

Anti-Ferroportin 1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Fornitore: Novus Biologicals
NBP1-21502PE
NOVUNBP1-21502PEEA 864 EUR
NOVUNBP1-21502PE
Anti-Ferroportin 1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Type: Primary
Antigen: Ferroportin 1
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR