Stampa…

Anti-THOP1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 4D6]

Fornitore: Novus Biologicals
NB400-146F
NOVUNB400-146FEA 655 EUR
NOVUNB400-146F
Anti-THOP1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 4D6]
Anticorpi
Type: Primary
Antigen: THOP1 (thimet oligopeptidase 1)
Clonality: monoclonal
Clone: 4D6
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR